Assay Details
Gene Symbol: GLY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111922.4 | NP_187696.1 | 4 - 5 | 642 | 79 | |
GenBank mRNA | AK228542.1 | - | 4 - 5 | 456 | 79 | |
AY052329.1 | - | 4 - 5 | 432 | 79 | ||
BT000849.1 | - | 4 - 5 | 403 | 79 | ||
Y08357.1 | - | 424 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
