Assay Details
Gene Symbol: GLY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111922.4 | NP_187696.1 | 6 - 7 | 937 | 104 | |
GenBank mRNA | AK228542.1 | - | 6 - 7 | 751 | 104 | |
AY052329.1 | - | 6 - 7 | 727 | 104 | ||
BT000849.1 | - | 6 - 7 | 698 | 104 | ||
Y08357.1 | - | 719 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
