Assay Details
Gene Symbol: PHS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113857.3 | NP_189578.1 | 13 - 14 | 2639 | 83 | |
GenBank mRNA | AY049235.1 | - | 13 - 14 | 2624 | 83 | |
BT003015.1 | - | 13 - 14 | 2575 | 83 | ||
KJ138807.1 | - | 13 - 14 | 2575 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
