Assay Details
Gene Symbol: MES11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339038.1 | NP_001327129.1 | 1 - 2 | 557 | 117 | |
NM_113902.4 | NP_189622.1 | 1 - 2 | 752 | 117 | ||
GenBank mRNA | BT003149.1 | - | 1 - 2 | 377 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
