Assay Details
Gene Symbol: AT3G45810 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114450.3 | NP_190167.2 | 9 - 10 | 2421 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
