Assay Details
Gene Symbol: AT3G49190 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114778.4 | NP_190488.1 | 4 - 5 | 997 | 92 | |
GenBank mRNA | AK117581.1 | - | 4 - 5 | 997 | 92 | |
BT005436.1 | - | 4 - 5 | 982 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
