Assay Details
Gene Symbol: AT3G49190 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114778.4 | NP_190488.1 | 6 - 7 | 1341 | 112 | |
GenBank mRNA | AK117581.1 | - | 6 - 7 | 1341 | 112 | |
BT005436.1 | - | 6 - 7 | 1326 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
