Assay Details
Gene Symbol: AT3G50400 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114900.3 | NP_190609.1 | 4 - 5 | 935 | 106 | |
GenBank mRNA | AK117654.1 | - | 4 - 5 | 936 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
