Assay Details
Gene Symbol: MES10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114904.4 | NP_566932.4 | 1 - 2 | 399 | 70 | |
GenBank mRNA | AY074858.1 | - | 1 - 2 | 400 | 70 | |
AY142031.1 | - | 1 - 2 | 399 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
