Assay Details
Gene Symbol: AT3G50990 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114959.3 | NP_190668.2 | 3 - 4 | 636 | 118 | |
GenBank mRNA | AK229843.1 | - | 3 - 4 | 636 | 118 | |
AK229896.1 | - | 3 - 4 | 636 | 118 | ||
AK229990.1 | - | 3 - 4 | 636 | 118 | ||
AK230084.1 | - | 3 - 4 | 636 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
