Assay Details
Gene Symbol: MES12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117058.4 | NP_192728.2 | 1 - 2 | 555 | 118 | |
GenBank mRNA | AK221404.1 | - | 1 - 2 | 396 | 118 | |
AY054619.1 | - | 1 - 2 | 467 | 118 | ||
AY081515.1 | - | 1 - 2 | 258 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
