Assay Details
Gene Symbol: MES12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117058.4 | NP_192728.2 | 2 - 3 | 798 | 77 | |
GenBank mRNA | AK221404.1 | - | 2 - 3 | 639 | 77 | |
AY054619.1 | - | 2 - 3 | 710 | 77 | ||
AY081515.1 | - | 2 - 3 | 501 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
