Assay Details
Gene Symbol: UGE5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117166.3 | NP_192834.1 | 6 - 7 | 856 | 72 | |
GenBank mRNA | AY065354.1 | - | 6 - 7 | 860 | 72 | |
AY117180.1 | - | 6 - 7 | 778 | 72 | ||
AY140073.1 | - | 6 - 7 | 860 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
