Assay Details
Gene Symbol: BGLU10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341886.1 | NP_001328392.1 | 11 - 12 | 1224 | 115 | |
NM_118921.4 | NP_567787.1 | 12 - 13 | 1456 | 115 | ||
GenBank mRNA | AY057518.1 | - | 12 - 13 | 1296 | 115 | |
BT002654.1 | - | 12 - 13 | 1286 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
