Assay Details
Gene Symbol: MYB41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118951.3 | NP_194540.1 | 2 - 3 | 401 | 92 | |
GenBank mRNA | AY519610.1 | - | 2 - 3 | 260 | 92 | |
BT001930.1 | - | 2 - 3 | 260 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
