Assay Details
Gene Symbol: AAE17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343787.1 | NP_001332667.1 | 5 - 6 | 1053 | 142 | |
NM_122211.3 | NP_197696.2 | 5 - 6 | 1053 | 142 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
