Assay Details
Gene Symbol: AT5G25590 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122470.3 | NP_197941.1 | 2 - 3 | 1501 | 134 | |
GenBank mRNA | AY091053.1 | - | 2 - 3 | 1358 | 134 | |
AY133786.1 | - | 2 - 3 | 1285 | 134 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
