Assay Details
Gene Symbol: AT5G25590 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122470.3 | NP_197941.1 | 3 - 4 | 1731 | 121 | |
GenBank mRNA | AY091053.1 | - | 3 - 4 | 1588 | 121 | |
AY133786.1 | - | 3 - 4 | 1515 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
