Assay Details
Gene Symbol: RCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102360.4 | NP_173920.1 | 6 - 7 | 1148 | 67 | |
GenBank mRNA | AY120757.1 | - | 6 - 7 | 1090 | 67 | |
BT000108.1 | - | 5 - 6 | 973 | 67 | ||
U21557.1 | - | 6 - 7 | 1051 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
