Assay Details
Gene Symbol: AT1G26470 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102410.5 | NP_564248.1 | 1 - 2 | 168 | 86 | |
GenBank mRNA | AY150388.1 | - | 1 - 2 | 125 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
