Assay Details
Gene Symbol: AT1G28690 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102636.2 | NP_174190.2 | 1 | 1496 | 145 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: AT1G28690 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102636.2 | NP_174190.2 | 1 | 1496 | 145 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |