Assay Details
Gene Symbol: PIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106017.4 | NP_177500.1 | 2 - 3 | 1584 | 93 | |
GenBank mRNA | AF372950.1 | - | 2 - 3 | 1584 | 93 | |
AY093960.1 | - | 2 - 3 | 1485 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
