Assay Details
Gene Symbol: PIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106017.4 | NP_177500.1 | 5 - 6 | 1903 | 97 | |
GenBank mRNA | AF372950.1 | - | 5 - 6 | 1903 | 97 | |
AY093960.1 | - | 5 - 6 | 1804 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
