Assay Details
Gene Symbol: AT1G73740 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106033.4 | NP_177515.1 | 1 - 2 | 1353 | 103 | |
GenBank mRNA | AY059826.1 | - | 1 - 2 | 1098 | 103 | |
AY064640.1 | - | 1 - 2 | 926 | 103 | ||
KJ138939.1 | - | 1 - 2 | 926 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
