Assay Details
Gene Symbol: AT1G73740 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106033.4 | NP_177515.1 | 2 - 3 | 1475 | 102 | |
GenBank mRNA | AY059826.1 | - | 2 - 3 | 1220 | 102 | |
AY064640.1 | - | 2 - 3 | 1048 | 102 | ||
KJ138939.1 | - | 2 - 3 | 1048 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
