Assay Details
Gene Symbol: AT5G27530 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122635.2 | NP_198105.2 | 7 - 8 | 886 | 193 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
