Assay Details
Gene Symbol: GRXC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123401.4 | NP_198853.1 | 1 - 2 | 255 | 69 | |
GenBank mRNA | AY064153.1 | - | 1 - 2 | 102 | 69 | |
AY072307.1 | - | 1 - 2 | 153 | 69 | ||
AY097411.1 | - | 1 - 2 | 51 | 69 | ||
AY114546.1 | - | 1 - 2 | 51 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
