Assay Details
Gene Symbol: GRXC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125867.1 | NP_001119339.1 | 2 - 3 | 615 | 89 | |
NM_123401.4 | NP_198853.1 | 3 - 4 | 446 | 89 | ||
GenBank mRNA | AY064153.1 | - | 3 - 4 | 293 | 89 | |
AY072307.1 | - | 3 - 4 | 344 | 89 | ||
AY086319.1 | - | 3 - 4 | 324 | 89 | ||
AY097411.1 | - | 3 - 4 | 242 | 89 | ||
AY114546.1 | - | 3 - 4 | 242 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
