Assay Details
Gene Symbol: PME5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124123.5 | NP_199561.1 | 1 - 2 | 343 | 123 | |
GenBank mRNA | AY102117.1 | - | 1 - 2 | 325 | 123 | |
BT000565.1 | - | 1 - 2 | 296 | 123 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
