Assay Details
Gene Symbol: NUDT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344736.1 | NP_001331651.1 | 2 - 3 | 748 | 84 | |
NM_001344737.1 | NP_001331652.1 | 3 - 4 | 429 | 84 | ||
NM_001344738.1 | NP_001331649.1 | 2 - 3 | 375 | 84 | ||
NM_001344739.1 | NP_001331650.1 | 2 - 3 | 308 | 84 | ||
NM_124139.4 | NP_568687.1 | 3 - 4 | 479 | 84 | ||
GenBank mRNA | AY042806.1 | - | 3 - 4 | 313 | 84 | |
AY064646.1 | - | 2 - 3 | 237 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
