Assay Details
Gene Symbol: NUDT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344736.1 | NP_001331651.1 | 7 - 8 | 1161 | 124 | |
NM_001344737.1 | NP_001331652.1 | 8 - 9 | 842 | 124 | ||
NM_001344738.1 | NP_001331649.1 | 7 - 8 | 788 | 124 | ||
NM_001344739.1 | NP_001331650.1 | 7 - 8 | 721 | 124 | ||
NM_124139.4 | NP_568687.1 | 8 - 9 | 892 | 124 | ||
GenBank mRNA | AY042806.1 | - | 8 - 9 | 726 | 124 | |
AY064646.1 | - | 7 - 8 | 650 | 124 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
