Assay Details
Gene Symbol: ELP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161296.1 | NP_001154768.1 | 13 - 14 | 2027 | 61 | |
NM_001344757.1 | NP_001318757.1 | 14 - 15 | 2068 | 61 | ||
NM_124183.4 | NP_199620.4 | 13 - 14 | 1893 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
