Assay Details
Gene Symbol: AT5G48140 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124188.4 | NP_199625.1 | 1 - 2 | 225 | 148 | |
GenBank mRNA | AK118684.1 | - | 1 - 2 | 223 | 148 | |
BT009687.1 | - | 1 - 2 | 109 | 148 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
