Assay Details
Gene Symbol: AT5G48140 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124188.4 | NP_199625.1 | 2 - 3 | 532 | 105 | |
GenBank mRNA | AK118684.1 | - | 2 - 3 | 530 | 105 | |
BT009687.1 | - | 2 - 3 | 416 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
