Assay Details
Gene Symbol: AT5G48140 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124188.4 | NP_199625.1 | 3 - 4 | 1048 | 64 | |
GenBank mRNA | AK118684.1 | - | 3 - 4 | 1046 | 64 | |
BT009687.1 | - | 3 - 4 | 932 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
