Assay Details
Gene Symbol: CPI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203575.1 | NP_001190504.1 | 2 - 3 | 323 | 75 | |
NM_124419.5 | NP_568727.1 | 2 - 3 | 348 | 75 | ||
GenBank mRNA | AF216756.1 | - | 2 - 3 | 166 | 75 | |
BT025804.1 | - | 1 - 2 | 134 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
