Assay Details
Gene Symbol: CPI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203575.1 | NP_001190504.1 | 3 - 4 | 415 | 74 | |
NM_001344891.1 | NP_001331972.1 | 2 - 3 | 690 | 74 | ||
NM_124419.5 | NP_568727.1 | 3 - 4 | 440 | 74 | ||
GenBank mRNA | AF216756.1 | - | 3 - 4 | 258 | 74 | |
BT025804.1 | - | 2 - 3 | 226 | 74 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001344891.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
