Assay Details
Gene Symbol: SDHAF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203580.1 | NP_001190509.1 | 2 - 3 | 255 | 92 | |
NM_001344916.1 | NP_001330999.1 | 2 - 3 | 402 | 92 | ||
NM_124483.3 | NP_199917.1 | 2 - 3 | 402 | 92 | ||
GenBank mRNA | AK229201.1 | - | 2 - 3 | 246 | 92 | |
BT025542.1 | - | 2 - 3 | 215 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
