Assay Details
Gene Symbol: SDHAF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036975.1 | NP_001032052.1 | 3 - 4 | 377 | 111 | |
NM_124483.3 | NP_199917.1 | 3 - 4 | 536 | 111 | ||
GenBank mRNA | AK229201.1 | - | 3 - 4 | 380 | 111 | |
AK317168.1 | - | 3 - 4 | 377 | 111 | ||
BT025542.1 | - | 3 - 4 | 349 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
