Assay Details
Gene Symbol: EMB3142 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344925.1 | NP_001318776.1 | 28 - 29 | 3464 | 107 | |
NM_001344926.1 | NP_001332447.1 | 28 - 29 | 3747 | 107 | ||
NM_001344927.1 | NP_001332446.1 | 28 - 29 | 3521 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
