Assay Details
Gene Symbol: SBT1.3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124554.3 | NP_568765.1 | 1 | 2655 | 108 | |
GenBank mRNA | AY080831.1 | - | 1 | 2462 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
