Assay Details
Gene Symbol: AT5G51890 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124568.6 | NP_200002.3 | 1 - 2 | 283 | 82 | |
GenBank mRNA | AY072121.1 | - | 1 - 2 | 274 | 82 | |
AY122968.1 | - | 1 - 2 | 208 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
