Assay Details
Gene Symbol: AT5G51890 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124568.6 | NP_200002.3 | 2 - 3 | 471 | 64 | |
GenBank mRNA | AY072121.1 | - | 2 - 3 | 462 | 64 | |
AY122968.1 | - | 2 - 3 | 396 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
