Assay Details
Gene Symbol: PLR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124738.4 | NP_200170.2 | 6 - 7 | 1009 | 71 | |
GenBank mRNA | AK221481.1 | - | 6 - 7 | 975 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
