Assay Details
Gene Symbol: E2F3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129160.4 | NP_565831.3 | 3 - 4 | 657 | 96 | |
NM_201882.3 | NP_973611.1 | 3 - 4 | 683 | 96 | ||
GenBank mRNA | AF242582.1 | - | 3 - 4 | 920 | 96 | |
AJ294534.1 | - | 3 - 4 | 511 | 96 | ||
BT026376.1 | - | 3 - 4 | 511 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
