Assay Details
Gene Symbol: E2F3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129160.4 | NP_565831.3 | 6 - 7 | 942 | 67 | |
NM_201882.3 | NP_973611.1 | 6 - 7 | 974 | 67 | ||
GenBank mRNA | AF242582.1 | - | 6 - 7 | 1205 | 67 | |
AJ294534.1 | - | 6 - 7 | 802 | 67 | ||
BT026376.1 | - | 6 - 7 | 802 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
