Assay Details
Gene Symbol: E2F3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129160.4 | NP_565831.3 | 9 - 10 | 1108 | 124 | |
NM_201881.2 | NP_973610.1 | 9 - 10 | 1154 | 124 | ||
NM_201882.3 | NP_973611.1 | 9 - 10 | 1140 | 124 | ||
GenBank mRNA | AF242582.1 | - | 9 - 10 | 1371 | 124 | |
AJ271597.1 | - | 9 - 10 | 1154 | 124 | ||
AJ294534.1 | - | 9 - 10 | 968 | 124 | ||
BT026376.1 | - | 9 - 10 | 968 | 124 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
