Assay Details
Gene Symbol: AT2G36370 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129194.4 | NP_565845.3 | 15 - 16 | 2466 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
