Assay Details
Gene Symbol: SBE2.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129196.4 | NP_181180.1 | 14 - 15 | 1700 | 77 | |
GenBank mRNA | AK226896.1 | - | 14 - 15 | 1651 | 77 | |
AY136411.1 | - | 14 - 15 | 1674 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
