Assay Details
Gene Symbol: AT2G37730 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129329.2 | NP_565869.1 | 3 - 4 | 1435 | 79 | |
GenBank mRNA | KJ138861.1 | - | 3 - 4 | 1435 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
